Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Omp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651575.1 | XP_007649765.1 | 1 | 1523 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |