Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496626.1 | XP_003496674.1 | 3 - 4 | 334 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |