Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497801.1 | XP_003497849.1 | 3 - 4 | 389 | 73 | |
XM_003497802.1 | XP_003497850.1 | 2 - 3 | 221 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |