Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509554.1 | XP_003509602.1 | 10 - 11 | 1459 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |