Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510823.2 | XP_003510871.1 | 4 - 5 | 847 | 67 | |
XM_007651405.1 | XP_007649595.1 | 4 - 5 | 768 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |