Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645024.1 | XP_007643214.1 | 13 - 14 | 3107 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |