Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500333.1 | XP_003500381.1 | 18 - 19 | 2658 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |