Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Artn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502690.2 | XP_003502738.1 | 3 - 4 | 508 | 90 | |
XM_007643282.1 | XP_007641472.1 | 4 - 5 | 658 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |