Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504176.1 | XP_003504224.1 | 1 | 722 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Both primers and probe map within a single exon |