Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508522.2 | XP_003508570.2 | 1 | 322 | 102 | |
GenBank mRNA | JI891197.1 | - | 1 - 2 | 76 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Both primers and probe map within a single exon |