Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508522.2 | XP_003508570.2 | 1 | 504 | 90 | |
GenBank mRNA | JI891197.1 | - | 2 - 3 | 258 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |