Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652836.1 | XP_007651026.1 | 6 - 7 | 792 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |