Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507493.2 | XP_003507541.1 | 2 - 3 | 1201 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |