Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510805.2 | XP_003510853.1 | 2 - 3 | 1091 | 76 | |
GenBank mRNA | JI865495.1 | - | 2 - 3 | 1096 | 76 | |
JP048075.1 | - | 2 - 3 | 1096 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |