Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513777.1 | XP_003513825.1 | 5 - 6 | 609 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |