Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650958.1 | XP_007649148.1 | 4 - 5 | 624 | 85 | |
GenBank mRNA | JI870208.1 | - | 1 - 2 | 236 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |