Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499165.2 | XP_003499213.1 | 1 - 2 | 72 | 63 | |
XM_003499166.2 | XP_003499214.1 | 1 - 2 | 57 | 63 | ||
XM_007638784.1 | XP_007636974.1 | 1 - 2 | 57 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |