Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499346.2 | XP_003499394.1 | 1 - 2 | 204 | 68 | |
GenBank mRNA | JI888525.1 | - | 1 - 2 | 204 | 68 | |
JP046220.1 | - | 1 - 2 | 204 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Endogenous Controls Recommended Assay Inventoried Probe spans exons |