Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100754798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643250.1 | XP_007641440.1 | 9 - 10 | 1272 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |