Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496043.2 | XP_003496091.1 | 1 - 2 | 257 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Probe spans exons |