Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499891.1 | XP_003499939.1 | 4 - 5 | 943 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Probe spans exons |