Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507654.2 | XP_003507702.2 | 1 - 2 | 1089 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |