Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511573.2 | XP_003511621.1 | 4 - 5 | 705 | 115 | |
GenBank mRNA | JI896737.1 | - | 3 - 4 | 729 | 115 | |
JP060486.1 | - | 3 - 4 | 729 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |