Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513776.1 | XP_003513824.1 | 4 - 5 | 376 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |