Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripply3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648608.1 | XP_007646798.1 | 6 - 7 | 800 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |