Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510327.1 | XP_003510375.1 | 5 - 6 | 1917 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |