Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511529.2 | XP_003511577.1 | 7 - 8 | 604 | 83 | |
GenBank mRNA | JP041294.1 | - | 4 - 5 | 381 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |