Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514409.1 | XP_003514457.1 | 1 - 2 | 60 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |