Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf12l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648456.1 | XP_007646646.1 | 1 - 2 | 1180 | 97 | |
GenBank mRNA | JP049948.1 | - | 1 - 2 | 979 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |