Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647168.1 | XP_007645358.1 | 2 - 3 | 168 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |