Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496071.2 | XP_003496119.1 | 2 - 3 | 451 | 95 | |
GenBank mRNA | JI872990.1 | - | 3 - 4 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |