Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501542.2 | XP_003501590.2 | 1 - 2 | 961 | 79 | |
XM_007642196.1 | XP_007640386.1 | 1 - 2 | 961 | 79 | ||
XM_007642198.1 | XP_007640388.1 | 1 - 2 | 961 | 79 | ||
XM_007642199.1 | XP_007640389.1 | 2 - 3 | 223 | 79 | ||
XM_007642200.1 | XP_007640390.1 | 2 - 3 | 169 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |