Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510737.1 | XP_003510785.1 | 3 - 4 | 711 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |