Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501169.2 | XP_003501217.1 | 2 - 3 | 194 | 141 | |
GenBank mRNA | JI878834.1 | - | 1 - 2 | 99 | 141 | |
JP041711.1 | - | 1 - 2 | 99 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |