Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100774508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509753.2 | XP_003509801.1 | 13 - 14 | 1767 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |