Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497857.1 | XP_003497905.1 | 6 - 7 | 845 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |