Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mansc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501259.2 | XP_003501307.1 | 3 - 4 | 803 | 63 | |
XM_007641925.1 | XP_007640115.1 | 4 - 5 | 958 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |