Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506161.1 | XP_003506209.1 | 4 - 5 | 614 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |