Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcwpw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648520.1 | XP_007646710.1 | 4 - 5 | 660 | 114 | |
XM_007648522.1 | XP_007646712.1 | 4 - 5 | 660 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |