Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lepre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653632.1 | XP_007651822.1 | 8 - 9 | 1139 | 103 | |
GenBank mRNA | JP053067.1 | - | 7 - 8 | 859 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |