Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514975.2 | XP_003515023.1 | 3 - 4 | 1119 | 71 | |
XM_007654988.1 | XP_007653178.1 | 4 - 5 | 674 | 71 | ||
XM_007654989.1 | XP_007653179.1 | 4 - 5 | 835 | 71 | ||
XM_007654990.1 | XP_007653180.1 | 4 - 5 | 1294 | 71 | ||
XM_007654991.1 | XP_007653181.1 | 4 - 5 | 755 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |