Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649345.1 | XP_007647535.1 | 2 - 3 | 150 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Probe spans exons |