Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643105.1 | XP_007641295.1 | 4 - 5 | 606 | 73 | |
GenBank mRNA | JI892950.1 | - | 4 - 5 | 606 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |