Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651253.1 | XP_007649443.1 | 3 - 4 | 516 | 100 | |
XM_007651254.1 | XP_007649444.1 | 3 - 4 | 516 | 100 | ||
GenBank mRNA | JI880477.1 | - | 1 - 2 | 109 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |