Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dytn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642720.1 | XP_007640910.1 | 6 - 7 | 552 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |