Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496979.2 | XP_003497027.1 | 3 - 4 | 551 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |