Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53i13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506088.2 | XP_003506136.1 | 2 - 3 | 165 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |