Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stoml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507370.2 | XP_003507418.1 | 3 - 4 | 230 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |