Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503378.2 | XP_003503426.2 | 2 - 3 | 282 | 80 | |
GenBank mRNA | JP052801.1 | - | 282 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Detect Genomic DNA Probe spans exons |