Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506496.1 | XP_003506544.1 | 1 - 2 | 137 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Probe spans exons |